Learn More
Cayman Chemical ADAMTS-5 Inhibitor, Purity: ≥95%, CAS: 929634-33-3, 1 mg
SDP
Catalog No. 502979998
Encompass_Preferred
An inhibitor of ADAMTS-5 (aggrecanase-2; IC50 = 1.1 µM) with 40-fold selectivity over ADAMTS-4 (aggrecanase-1)
Catalog No. 50-297-9998 Supplier Cayman Chemical Supplier No. 171881 MG
An inhibitor of ADAMTS-5 (aggrecanase-2; IC50 = 1.1 µM) with 40-fold selectivity over ADAMTS-4 (aggrecanase-1)