Learn More
Medchemexpress LLC ADAMTS13 Human CHO 50ug | 50ug
SDP
Catalog No. 5000194797
Encompass_Preferred
The ADAMTS13 protein a metalloproteinase has a metalloproteinase domain a disintegrin-like domain and a thrombospondin type 1 motif It cleaves von Willebrand Factor and defects in this gene cause thrombotic thrombocytopenic purpura Alternative splicing generates multiple transcript variants ADAMTS13 is broadly expressed in various tissues including the liver testis and 24 other tissues ADAMTS13 Protein Human (CHO His) is the recombinant human-derived ADAMTS13 expressed by CHO with C-10 His labeled tag The total length of ADAMTS13 Protein Human (CHO His) is 655 a a
Catalog No. 50-001-94797 Supplier Medchemexpress LLC Supplier No. HYP7914450UG
The ADAMTS13 protein a metalloproteinase has a metalloproteinase domain a disintegrin-like domain and a thrombospondin type 1 motif It cleaves von Willebrand Factor and defects in this gene cause thrombotic thrombocytopenic purpura Alternative splicing generates multiple transcript variants ADAMTS13 is broadly expressed in various tissues including the liver testis and 24 other tissues ADAMTS13 Protein Human (CHO His) is the recombinant human-derived ADAMTS13 expressed by CHO with C-10 His labeled tag The total length of ADAMTS13 Protein Human (CHO His) is 655 a a