Learn More
AnaSpec ADAMTS-4/Aggrecanase FRET Substrate, FRET Assay, 1 mg
Catalog No. NC3451015
Encompass
The ADAMTS-4/Aggrecanase FRET Substrate offers researchers a reliable tool for detecting ADAMTS-4 enzyme activity. With a quantity of 1 mg, this peptide substrate enables precise enzyme assays with FRET technology, distinguishing it from standard substrates. Its specific function in cartilage degradation pathways makes it vital for studies related to osteoarthritis and tissue remodeling. Easy to use and store, it supports ongoing research in enzyme substrates and inhibitors.
Catalog No. NC3451015 Supplier AnaSpec Supplier No. AS604311
The ADAMTS-4/Aggrecanase FRET Substrate offers researchers a reliable tool for detecting ADAMTS-4 enzyme activity. With a quantity of 1 mg, this peptide substrate enables precise enzyme assays with FRET technology, distinguishing it from standard substrates. Its specific function in cartilage degradation pathways makes it vital for studies related to osteoarthritis and tissue remodeling. Easy to use and store, it supports ongoing research in enzyme substrates and inhibitors.