Learn More
Aviva Systems Biology Corp ADAMTS4 Antibody - N-terminal region (ARP41556_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41556_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADAMTS4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ADAMTS4. Alternate names: ADAMTS-2, ADAMTS-4, ADMP-1, KIAA0688ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57904