Learn More
ProSci ADAMTS8 Peptide

Description
Specifications
Specifications
| Gene ID (Entrez) | 11095 |
| Species | Not Available |
| Accession Number | NP_008968.4 |
| Common Name | ADAM metallopeptidase with thrombospondin type 1 motif, 8 |
| Content And Storage | Store at -20°C. When reconstituted minimize freezing and thawing. |
| Format | Lyophilized |
| Formulation | ADAMTS8 peptide is supplied as 100 μg of dried peptide. When the peptide is reconstituted in 200 μL water, the concentration will be 0.5 mg/mL |
| For Use With (Application) | Blocking Assay |
| Gene Symbol | ADAMTS8 |
| Product Type | Blocking Peptide |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.