Learn More
Chemglass Life Sciences Adapter, 1-1/2" Beaded Pipe to 1" Sanitary, PTFE
SDP
Catalog No. 501656786
Encompass_Preferred
Adapter, 1-1/2" Beaded Pipe to 1" Sanitary, PTFE. Beaded pipe to sanitary fitting adapter connects to the 1-1/2" beaded pipe side arm of CG-1968-GL zero dead space drain valve. Approximate overall length of adapter is 1.75". NOTE: Requires a CG-1968 beaded pipe coupling.
Catalog No. 50-165-6786 Supplier Chemglass Life Sciences Supplier No. CG1964B150
Adapter, 1-1/2" Beaded Pipe to 1" Sanitary, PTFE. Beaded pipe to sanitary fitting adapter connects to the 1-1/2" beaded pipe side arm of CG-1968-GL zero dead space drain valve. Approximate overall length of adapter is 1.75". NOTE: Requires a CG-1968 beaded pipe coupling.