Learn More
Julabo USA Inc Pair, 10mm hose barb adapters
SDP
Catalog No. 507534220
Encompass_Preferred
Bath Accessories, Julabo, Adapter barbed, for 10mm ID tubing, pair (for CD/CP/DD/MS/MX, HE/HL, SE/SL). Requires M16x1 female nut to install.
Catalog No. 50-753-4220 Supplier Julabo USA Inc Supplier No. 8970447
Bath Accessories, Julabo, Adapter barbed, for 10mm ID tubing, pair (for CD/CP/DD/MS/MX, HE/HL, SE/SL). Requires M16x1 female nut to install.