Learn More
Julabo USA Inc Pair, 3/4" FNPT to 1/2" hose barb adapters
SDP
Catalog No. 507534196
Encompass_Preferred
Bath Accessories, Julabo, Adapter barbed for 1/2 in. diameter tubing to NPT 3/4 in. female, pair
Catalog No. 50-753-4196 Supplier Julabo USA Inc Supplier No. 8890036
Bath Accessories, Julabo, Adapter barbed for 1/2 in. diameter tubing to NPT 3/4 in. female, pair