Learn More
Julabo USA Inc Pair, M10x1m to 12mm hose barb adapters
SDP
Catalog No. 507534222
Encompass_Preferred
Bath Accessories, Julabo, Adapter barbed, 13.5mm OD to M10x1 male., pair (for F250, AWC100). For use with 12mm (1/2") ID rubber tubing.
Catalog No. 50-753-4222 Supplier Julabo USA Inc Supplier No. 8970468
Bath Accessories, Julabo, Adapter barbed, 13.5mm OD to M10x1 male., pair (for F250, AWC100). For use with 12mm (1/2") ID rubber tubing.