Learn More
Julabo USA Inc Pair, M10x1m to 8mm hose barb adapters
SDP
Catalog No. 507534221
Encompass_Preferred
Bath Accessories, Julabo, Adapter barbed, 9.5mm OD to M10x1 male., pair (for ED, EH, MB, MC, ME, F250, AWC100). For use with 8mm ID rubber tubing.
Catalog No. 50-753-4221 Supplier Julabo USA Inc Supplier No. 8970460
Bath Accessories, Julabo, Adapter barbed, 9.5mm OD to M10x1 male., pair (for ED, EH, MB, MC, ME, F250, AWC100). For use with 8mm ID rubber tubing.