Learn More
Ace Glass, Inc. Column end fitting, #11 to 1/4-28 UNF helicoil tap, paper filter disc not included
SDP
Catalog No. 50978437
Encompass_Preferred
#11 Ace-Thred column end fitting adapter with -28UNF tap with 18-8 stainless helicoil insert for durability. Design allows for a leak-tight seal without the need for an o-ring.
Catalog No. 50-978-437 Supplier Ace Glass, Inc. Supplier No. 580147
#11 Ace-Thred column end fitting adapter with -28UNF tap with 18-8 stainless helicoil insert for durability. Design allows for a leak-tight seal without the need for an o-ring.