Learn More
Julabo USA Inc Pair, G 1 1/4" female to 1" hose barb adapters
SDP
Catalog No. 507534205
Encompass_Preferred
Bath Accessories, Julabo, Adapters G 1 1/4" female to barbed fittings for tubing 1" inner dia. for FL(W)2506/4006/7006/11006/20006
Catalog No. 50-753-4205 Supplier Julabo USA Inc Supplier No. 8890046
Bath Accessories, Julabo, Adapters G 1 1/4" female to barbed fittings for tubing 1" inner dia. for FL(W)2506/4006/7006/11006/20006