Learn More
Julabo USA Inc Pair, G 1 1/4" female to 1/2" MNPT adapters
SDP
Catalog No. 507534208
Encompass_Preferred
Bath Accessories, Julabo, Adapters G 1 1/4" female to NPT 1/2" male for FL(W)2506/4006/7006/11006/20006
Catalog No. 50-753-4208 Supplier Julabo USA Inc Supplier No. 8890049
Bath Accessories, Julabo, Adapters G 1 1/4" female to NPT 1/2" male for FL(W)2506/4006/7006/11006/20006