Learn More
Julabo USA Inc Pair, G 3/4" female to 1/2" hose barb adapters
SDP
Catalog No. 507534201
Encompass_Preferred
Bath Accessories, Julabo, Adapters G 3/4" female to barbed fittings for tubing 1/2" inner diameter for FL(W)1203/1703/2503/4003
Catalog No. 50-753-4201 Supplier Julabo USA Inc Supplier No. 8890042
Bath Accessories, Julabo, Adapters G 3/4" female to barbed fittings for tubing 1/2" inner diameter for FL(W)1203/1703/2503/4003