Learn More
Cayman Chemical Adapalene-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1276433-89-6, 1 mg
SDP
Catalog No. 502933121
Encompass_Preferred
An internal standard for the quantification of adapalene by GC- or LC-MS
Catalog No. 50-293-3121 Supplier Cayman Chemical Supplier No. 257581 MG
An internal standard for the quantification of adapalene by GC- or LC-MS