Learn More
Cayman Chemical Adapalen-d3 500ug
SDP
Catalog No. 502933122
Encompass_Preferred
An internal standard for the quantification of adapalene by GC- or LC-MS
Catalog No. 50-293-3122 Supplier Cayman Chemical Supplier No. 25758500 UG
Add to Cart
Add to Cart
An internal standard for the quantification of adapalene by GC- or LC-MS