Learn More
Apexbio Technology LLC Adapalene 106685-40-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000096602
Encompass_Preferred
Adapalene (106685-40-9) is a small-molecule agonist targeting retinoic acid receptor subtypes RAR and RAR with lower affinity for RAR and minimal activity toward RXR It is designed to selectively modulate retinoic acid receptor-mediated gene transcription thereby regulating cellular growth differentiation and apoptosis Adapalene exerts its biological activity primarily through selective activation of RAR and RAR subtypes In experimental models adapalene exhibits agonistic activity with Ki values of 2 3 nM for RAR 9 3 nM for RAR and 22 nM for RAR Based on these pharmacological properties adapalene holds research potential in investigating retinoid signaling pathways cellular proliferation apoptosis and antitumor mechanisms particularly in colorectal cancer models
Catalog No. 50-000-96602 Supplier Apexbio Technology LLC Supplier No. A12675.1
Adapalene (106685-40-9) is a small-molecule agonist targeting retinoic acid receptor subtypes RAR and RAR with lower affinity for RAR and minimal activity toward RXR It is designed to selectively modulate retinoic acid receptor-mediated gene transcription thereby regulating cellular growth differentiation and apoptosis Adapalene exerts its biological activity primarily through selective activation of RAR and RAR subtypes In experimental models adapalene exhibits agonistic activity with Ki values of 2 3 nM for RAR 9 3 nM for RAR and 22 nM for RAR Based on these pharmacological properties adapalene holds research potential in investigating retinoid signaling pathways cellular proliferation apoptosis and antitumor mechanisms particularly in colorectal cancer models