Learn More
Enzo Life Sciences Adapalene (20mg). CAS: 106685-40-9
SDP
Supplier:  Enzo Life Sciences BMLGR1110020
Encompass_Preferred
Adapalene is a stable analog of retinoic acid and acts selectively as an agonist at RARβ and RARγ. It inhibits proliferation and induces apoptosis in colorectal cancer cells. Its unique pharmacological properties make it superior to other retinoids for the treatment of acne. Alternative name: 6-[3-(1-Adamantyl)-4-methoxyphenyl]-2-naphthoic acid. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (3 mg/mL) or DMF (6 mg/mL). Long Term Storage: Ambient.
Catalog No. 50-201-0924