Learn More
Selleck Chemical LLC Adapalene S1276-25mg
SDP
Catalog No. 503108260
Encompass_Preferred
Adapalene (CD-271) is a dual RAR and RXR agonist used in the treatment of acne
Catalog No. 50-310-8260 Supplier Selleck Chemical LLC Supplier No. S127625MG
Adapalene (CD-271) is a dual RAR and RXR agonist used in the treatment of acne