Learn More
Selleck Chemical LLC Adapalene S1276-100mg
SDP
Catalog No. 503110667
Encompass_Preferred
Adapalene (CD-271) is a dual RAR and RXR agonist used in the treatment of acne
Catalog No. 50-311-0667 Supplier Selleck Chemical LLC Supplier No. S1276100MG
Adapalene (CD-271) is a dual RAR and RXR agonist used in the treatment of acne