Learn More
TARGETMOL CHEMICALS INC Adapalene 500MG
SDP
Catalog No. 502877637
Encompass_Preferred
Also available in 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Adapalene (CD271) is a dual RAR and RXR agonist, utilized in the treatment of skin conditions. Purity 99.88%
Catalog No. 50-287-7637 Supplier TARGETMOL CHEMICALS INC Supplier No. T1093500MG
Also available in 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Adapalene (CD271) is a dual RAR and RXR agonist, utilized in the treatment of skin conditions. Purity 99.88%