Learn More
Medchemexpress LLC Adapalene (CD271) | 106685-40-9 | 99.9% | C28H28O3 | 500 MG
SDP
Catalog No. 5000432805
Encompass_Preferred
Adapalene (CD271) is a third-generation synthetic retinoid primarily used for acne research. It functions as a potent RAR agonist and non-competitively inhibits GOT1 enzymatic activity, also demonstrating anti-tumor properties.
  • Acts as a potent RAR agonist with varying AC50s for RAR subtypes.
  • Inhibits viability of various cancer cell lines.
  • Induces apoptosis and inhibits proliferation in ES-2 cells.
  • Increases G1-phase population in LoVo or DLD1 cells.
  • Inhibits GOT1 activity.
  • Inhibits expression of plasma membrane-associated enzyme transglutaminase Type I.
  • Inhibits growth of DLD1 cell-derived xenograft tumors in BALB/C nude mice, reducing tumor weight and volume.
Catalog No. 50-004-32805 Supplier Medchemexpress LLC Supplier No. HYB0091500MG
Adapalene (CD271) is a third-generation synthetic retinoid primarily used for acne research. It functions as a potent RAR agonist and non-competitively inhibits GOT1 enzymatic activity, also demonstrating anti-tumor properties.
  • Acts as a potent RAR agonist with varying AC50s for RAR subtypes.
  • Inhibits viability of various cancer cell lines.
  • Induces apoptosis and inhibits proliferation in ES-2 cells.
  • Increases G1-phase population in LoVo or DLD1 cells.
  • Inhibits GOT1 activity.
  • Inhibits expression of plasma membrane-associated enzyme transglutaminase Type I.
  • Inhibits growth of DLD1 cell-derived xenograft tumors in BALB/C nude mice, reducing tumor weight and volume.