Learn More
Apexbio Technology LLC Adapalene sodium salt 911110-93-5 100mg
SDP
Supplier:  Apexbio Technology LLC B1277100
Encompass_Preferred
Adapalene sodium salt (CAS 911110-93-5) is a small-molecule agonist targeting retinoic acid receptors (RARs) It is designed to selectively modulate RAR-mediated signaling thereby regulating cellular differentiation and proliferation Adapalene sodium salt exerts its biological activity primarily through specific binding and activation of nuclear RAR subtypes influencing gene transcription It demonstrates agonist activity toward RARs with EC50 values of 2 3 nM 9 3 nM and 22 nM for different RAR subtypes Additionally Adapalene sodium salt noncompetitively inhibits an unspecified enzyme s activity Based on these pharmacological properties Adapalene sodium salt holds research potential in studies of retinoid signaling cellular differentiation mechanisms dermatological disease models and antitumor investigations
Catalog No. 50-000-92168