Learn More
Abcam Adaphostin, 25MG
Catalog No. 502680451
Encompass_Preferred
MW 393.5 Da, Purity >=98%. Adaphostin is a p210bcr/abl tyrosine kinase inhibitor (IC50 = 14 μM) that displays anti-proliferative activity in leukemia models, human prostate cancer cell line PC-3 and other cancer cell lines. Adaphostin is a potent inducer of myeloid cell death.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-0451 Supplier Abcam Supplier No. AB28713625MG
MW 393.5 Da, Purity >=98%. Adaphostin is a p210bcr/abl tyrosine kinase inhibitor (IC50 = 14 μM) that displays anti-proliferative activity in leukemia models, human prostate cancer cell line PC-3 and other cancer cell lines. Adaphostin is a potent inducer of myeloid cell death.
The product is subject to the following: Abcam Restricted Use Statement