Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex Fitting, Nylon, Straight, Hose Barb to Threaded Adapter, 3/8"" ID x 1/4"" NPT(M); 10/PK
Catalog No. 50628922
Encompass
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
Catalog No. 50-628-922 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX3061518
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.