Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hanna Pharmaceutical Supply Co., Inc. ADAPTACAP BOTTLE 28MM LONG NEC

Supplier:  Hanna Pharmaceutical Supply Co., Inc. H9385106

Encompass_Preferred

AdaptaCap Bottle Adapter, 28 mm, long neck
Please note: This item is nonrefundable, non-returnable and non-cancelable once order is placed.

Catalog No. NC2073287


May include imposed supplier surcharges.
Only null left
Add to Cart
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.