Learn More
Chemglass Life Sciences Adapter, Distillation, Vent, 0-14mm Valve with Inlet Valve and 45/50 Lower Inner Jt
SDP
Catalog No. NC1674304
Encompass_Preferred
Adapter, Distillation, Vent, 0-14mm Valve with Inlet Valve and 45/50 Lower Inner Jt. Part of the CG-1965-D distallation glassware kit. ADAPTER ONLY.
Catalog No. NC1674304 Supplier Chemglass Life Sciences Supplier No. CG1965D215
Adapter, Distillation, Vent, 0-14mm Valve with Inlet Valve and 45/50 Lower Inner Jt. Part of the CG-1965-D distallation glassware kit. ADAPTER ONLY.