Learn More
Chemglass Life Sciences Adapter 1-1/2" beaded pipe to M16 x 1 Male
SDP
Supplier:  Chemglass Life Sciences CG1969A08
Encompass_Preferred
Adapter 1-1/2" beaded pipe to M16 x 1 Male. Stainless steel transition adapter from beaded pipe on the inlet/outlet of the reactor to M16 x 1 or M30 x 1.5 male. Used to connect fluid transfer hose to the vessel. Adapters require the beaded pipe couplings for connection to the vessel. Two are required per reactor.
Catalog No. CG1969A08