Learn More
Med Vet International Adapter 1/16" to 3/32", J0484MD6
Catalog No. 5000581512
Encompass_Preferred
This allows smaller electrodes (1/16") to be used with Bovie handpieces (3/32"). Especially useful for smaller Ellman electrodes.
Catalog No. 50-005-81512 Supplier Med Vet International Supplier No. JOR484MD6
This allows smaller electrodes (1/16") to be used with Bovie handpieces (3/32"). Especially useful for smaller Ellman electrodes.