Learn More
IDEX Health & Science Upchurch ADAPTER 1/4-28 TO 5/16-24
Catalog No. NC1919745
Encompass_Preferred
Idex Adapters for HPLC, Connections: Female to 5/16-24 female, For Use With: 1/4-28 Flat Bottom Female to 5/16-24 Female Flat Bottom, PEEK, White, Diameter Inner: 0.08 in., Pressure Rating: 1000 psi, Swept Volume: 4.12 uL
Catalog No. NC1919745 Supplier IDEX Health & Science Upchurch Supplier No. P135
Idex Adapters for HPLC, Connections: Female to 5/16-24 female, For Use With: 1/4-28 Flat Bottom Female to 5/16-24 Female Flat Bottom, PEEK, White, Diameter Inner: 0.08 in., Pressure Rating: 1000 psi, Swept Volume: 4.12 uL