Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Adapter with 1/4 in. NPT Thread x 1/8 in. Barb - Natural Kynar
SDP

Catalog No. 01000035 Shop All Fisherbrand Products
Catalog No. 01-000-035

missing translation for 'validMainframeMsg'

Superior single barb technology that provides a leak-proof seal. Ideal with flexible tubing.

  • Single & Multi Use Applications
  • PVC-Free
  • Superior Single Barb Design
  • USP Class VI Compliant
  • USP 661
  • USP81
  • USP 8
  • RoHS Compliant
  • Gamma Stable
  • Animal Derivative Free
  • Lot Traceable

Warranty and Services

Replacement if not satisfied. 12 Month return Policy.

Specifications

Certifications/Compliance Compliance Certificates Available
Material Natural Kynar
For Use With (Application) Threaded Tube Connector for Medical, Pharmaceutical, Industrial applications.
Packaging Quantity 10 Pack
Storage Requirements Out of direct sunlight, >20°F <100°F
Color Natural
Product Type Threaded Adapter
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.