Learn More
Cole-Parmer Spex VapLock™ Fitting, Polypropylene, Straight, Hose Barb to Threaded Adapter, 3/8" ID x 1/4" NPT(M); 10/PK
Supplier:  Cole-Parmer 1202022
Encompass_Preferred
Adapters for flexible tubing let you connect 1/16" to 3/4" ID soft-wall tubing to NPT(F) ports. Adapters are made of polypropylene. Wrap NPT threads 3 to 4 times with PTFE tape and wrench tighten for leak-resistant connections.
Catalog No. NC2119834