Learn More
Julabo USA Inc Pair, M16x1f to 1/4" tube stub adapters
SDP
Catalog No. 507534190
Encompass_Preferred
Bath Accessories, Julabo, Adapter 1/4 in. tube, Pair of M16x1 female to 1/4" tube stub adapters. For use with Swagelok union fittings.
Catalog No. 50-753-4190 Supplier Julabo USA Inc Supplier No. 8890011
Bath Accessories, Julabo, Adapter 1/4 in. tube, Pair of M16x1 female to 1/4" tube stub adapters. For use with Swagelok union fittings.