Learn More
Chemglass Life Sciences Adapter, 1" Beaded Pipe to 3/4" Compression Fitting, Glass Filled TFE
SDP
Supplier:  Chemglass Life Sciences CG196450
Encompass_Preferred
Adapter, PTFE, 1" Beaded Pipe to 3/4" Tube. Glass filled PTFE adapter for use with the CG-1968-P zero dead space detachable valve. Adapts 1" beaded pipe to a 3/4" compression fitting for use with CG-1975 adapters and tubing. Requires a CG-1968-66 1" beaded pipe coupling.
Catalog No. CG196450