Learn More
Chemglass Life Sciences Adapter 1" beaded pipe to M16 x 1 Male
SDP
Catalog No. CG1969A05
Encompass_Preferred
Adapter 1" beaded pipe to M16 x 1 Male. Stainless steel transition adapter from beaded pipe on the inlet/outlet of the reactor to M16 x 1 or M30 x 1.5 male. Used to connect fluid transfer hose to the vessel. Adapters require the beaded pipe couplings for connection to the vessel. Two are required per reactor.
Catalog No. CG1969A05 Supplier Chemglass Life Sciences Supplier No. CG1969A05
Adapter 1" beaded pipe to M16 x 1 Male. Stainless steel transition adapter from beaded pipe on the inlet/outlet of the reactor to M16 x 1 or M30 x 1.5 male. Used to connect fluid transfer hose to the vessel. Adapters require the beaded pipe couplings for connection to the vessel. Two are required per reactor.