Learn More
Grainger Adapter 1 in CTS PEX White
Catalog No. 502763916
Encompass_Preferred
The Grainger 16T733 is a 3/4 HP, 115V, 12.8A, 60Hz, 1 phase, TEFC electric motor with a C-face mounting, 56C frame size, and 1.15 service factor. It has a RPM of 1725 at full load and is thermally protected. The motor is designed for continuous duty applications and features a cast iron frame and end bells along with ball bearings for smooth operation. With its powerful 3/4 HP rating, this electric motor is well-suited for driving pumps, fans, conveyors, and other industrial equipment.
Catalog No. 50-276-3916 Supplier Grainger Supplier No. 16T733
The Grainger 16T733 is a 3/4 HP, 115V, 12.8A, 60Hz, 1 phase, TEFC electric motor with a C-face mounting, 56C frame size, and 1.15 service factor. It has a RPM of 1725 at full load and is thermally protected. The motor is designed for continuous duty applications and features a cast iron frame and end bells along with ball bearings for smooth operation. With its powerful 3/4 HP rating, this electric motor is well-suited for driving pumps, fans, conveyors, and other industrial equipment.