Learn More
AVANTOR MASTERFLEX (VWR LLC) ADAPTER 10-32X1/16IN 250/PK
Catalog No. 502742275
Encompass
Masterflex Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 1/16IN ID×10-32 UNF(M); 250/PK, Avantor
Catalog No. 50-274-2275 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4062503
Masterflex Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 1/16IN ID×10-32 UNF(M); 250/PK, Avantor