Learn More
AVANTOR MASTERFLEX (VWR LLC) ADAPTER 10-32X3/32IN 250/PK
Catalog No. 502742277
Encompass
Masterflex Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 3/32IN ID x 10-32 UNF(M); 250/PK, Avantor
Catalog No. 50-274-2277 Supplier AVANTOR MASTERFLEX (VWR LLC) Supplier No. MFLX4062500
Masterflex Fitting, Nylon, Straight, Hosebarb to Thread Adapter, 3/32IN ID x 10-32 UNF(M); 250/PK, Avantor