Learn More
Beckman Coulter 17mm Diameter Polypropylene Tube Adapter Assembly
Catalog No. NC1617236
Encompass
Adapter for Tubes in Beckman Coulter Rotors used in High Performance and High Capacity Rotors: JS-5.3 AllSpin
Catalog No. NC1617236 Supplier Beckman Coulter Supplier No. 392075
Adapter for Tubes in Beckman Coulter Rotors used in High Performance and High Capacity Rotors: JS-5.3 AllSpin