Learn More
Chemglass Life Sciences Adapter, 18/9 Spherical Socket to 10 mm Hose Barb for Schlenk Line
SDP
Catalog No. NC3835827
Encompass_Preferred
This glass adapter provides a secure transition from an 18/9 spherical socket joint to a 10 mm hose barb, enabling Schlenk-style inert or vacuum manifolds to connect to flexible tubing for laboratory vacuum and inert line applications.
  • Connects 18/9 spherical socket joints to 10 mm hose.
  • Enables connection of Schlenk-style inert and vacuum lines to flexible tubing.
  • Provides a secure glass-to-hose seal suitable for vacuum and inert atmospheres.
  • Sold individually for easy replacement and maintenance.
Catalog No. NC3835827 Supplier Chemglass Life Sciences Supplier No. AF045409
This glass adapter provides a secure transition from an 18/9 spherical socket joint to a 10 mm hose barb, enabling Schlenk-style inert or vacuum manifolds to connect to flexible tubing for laboratory vacuum and inert line applications.
  • Connects 18/9 spherical socket joints to 10 mm hose.
  • Enables connection of Schlenk-style inert and vacuum lines to flexible tubing.
  • Provides a secure glass-to-hose seal suitable for vacuum and inert atmospheres.
  • Sold individually for easy replacement and maintenance.