Learn More
Chemglass Life Sciences Adapter, 1" Beaded Pipe to 1" Sanitary, 316L Stainless Steel
SDP
Catalog No. 501311167
Encompass_Preferred
Adapter, 1" Beaded Pipe to 1" Sanitary, 316L Stainless Steel Beaded pipe to sanitary fitting adapter connects to the 1¿ beaded pipe side arm of CG-1968-Q zero dead space drain valve. Approximate overall length of adapter is 2¿(52mm). NOTE: Requires a CG-1968-66 beaded coupling.
Catalog No. 50-131-1167 Supplier Chemglass Life Sciences Supplier No. CG1964B22
Adapter, 1" Beaded Pipe to 1" Sanitary, 316L Stainless Steel Beaded pipe to sanitary fitting adapter connects to the 1¿ beaded pipe side arm of CG-1968-Q zero dead space drain valve. Approximate overall length of adapter is 2¿(52mm). NOTE: Requires a CG-1968-66 beaded coupling.