Learn More
Chemglass Life Sciences Adapter, 1" Beaded Pipe to 1" Male NPT, Glass Filled TFE
SDP
Catalog No. 501531310
Encompass_Preferred
Adapter, 1" Beaded Pipe to 1" Male NPT Made from Glass Filled TFE. Glass filled PTFE adapter for use with side outlet of the CG-1949-F / CG-1959-F Filter Reactor Base or the CG-1968-Q zero dead space detachable valve. Adapts 1" beaded pipe to a Male NPT. Requires a CG-1968-66, 1" beaded pipe coupling.
Catalog No. 50-153-1310 Supplier Chemglass Life Sciences Supplier No. CG1964A12
Adapter, 1" Beaded Pipe to 1" Male NPT Made from Glass Filled TFE. Glass filled PTFE adapter for use with side outlet of the CG-1949-F / CG-1959-F Filter Reactor Base or the CG-1968-Q zero dead space detachable valve. Adapts 1" beaded pipe to a Male NPT. Requires a CG-1968-66, 1" beaded pipe coupling.