Learn More
Chemglass Life Sciences Adapter, Outer to 20-400 GPI Thread, Vacuum, Stopcock
SDP
Catalog No. 501214824
Encompass_Preferred
Adapter, Outer to 20-400 GPI Thread, Vacuum, Stopcock. Vacuum Adapter has a ground glass standard taper outer joint on top that provides a convenient leak free way to connect to the OP-6602, OP-6603 and OP-6604 disposable long stem filter funnels and a taper inner joint or a vial. The unique design of the long stem filter funnel and vacuum adapter makes it unnecessary to clean after each use. The 2mm PTFE stopcock between the outer joint and standard taper inner permits control of the vacuum source. Hose connection is 10mm OD at the largest serration. U.S. patent pending.
Catalog No. 50-121-4824 Supplier Chemglass Life Sciences Supplier No. CG1049J30
Adapter, Outer to 20-400 GPI Thread, Vacuum, Stopcock. Vacuum Adapter has a ground glass standard taper outer joint on top that provides a convenient leak free way to connect to the OP-6602, OP-6603 and OP-6604 disposable long stem filter funnels and a taper inner joint or a vial. The unique design of the long stem filter funnel and vacuum adapter makes it unnecessary to clean after each use. The 2mm PTFE stopcock between the outer joint and standard taper inner permits control of the vacuum source. Hose connection is 10mm OD at the largest serration. U.S. patent pending.