Learn More
Chemglass Life Sciences Adapter, 20-400 GPI Thread to 20-400 GPI Thread
SDP
Catalog No. 50872881
Encompass_Preferred
Adapter, Vacuum Take-Off, Long Stem, 24/40 Joint Size. With standard taper top outer and lower inner joints with sealed in lower tube extending below inner joint. Tubing extends 200mm below the joint. Serrated hose connection having an O.D. of 10mm at the largest serration for connection to vacuum source.
Catalog No. 50-872-881 Supplier Chemglass Life Sciences Supplier No. CG1049C06
Adapter, Vacuum Take-Off, Long Stem, 24/40 Joint Size. With standard taper top outer and lower inner joints with sealed in lower tube extending below inner joint. Tubing extends 200mm below the joint. Serrated hose connection having an O.D. of 10mm at the largest serration for connection to vacuum source.