Learn More
Chemglass Life Sciences Adapter, #20 O-Ring Joint to 1" Tubulation, Aifree, Schlenk
SDP
Catalog No. 501530950
Encompass_Preferred
Adapter, #20 O-Ring Joint to 1" Tubulation, Aifree, Schlenk. Lower adapter has a #20 o-ring joint with a 1" OD tubulation for connection to flexible vacuum tubing.For use with the AF-0300-50 drying tower. Image shown is full assembled tower.
Catalog No. 50-153-0950 Supplier Chemglass Life Sciences Supplier No. AF030051
Adapter, #20 O-Ring Joint to 1" Tubulation, Aifree, Schlenk. Lower adapter has a #20 o-ring joint with a 1" OD tubulation for connection to flexible vacuum tubing.For use with the AF-0300-50 drying tower. Image shown is full assembled tower.