Learn More
Chemglass Life Sciences Adapter, Outer to 24/25 Inner, Vacuum, Stopcock
SDP
Supplier:  Chemglass Life Sciences CG1049J24
Encompass_Preferred
Adapter, Outer to 24/25 Inner, Vacuum, Stopcock. Vacuum Adapter has a ground glass standard taper outer joint on top that provides a convenient leak free way to connect to the OP-6602, OP-6603 and OP-6604 disposable long stem filter funnels and a taper inner joint or a vial. The unique design of the long stem filter funnel and vacuum adapter makes it unnecessary to clean after each use. The 2mm PTFE stopcock between the outer joint and standard taper inner permits control of the vacuum source. Hose connection is 10mm OD at the largest serration. U.S. patent pending.
Catalog No. 50-121-4822