Learn More
Chemglass Life Sciences 1/8" Thermocouple Adapter, 24/40 Inner
SDP
Catalog No. CG1042E01
Encompass_Preferred
1/8" Thermocouple Adapter, 24/40 Inner. Adapter has a Chem-Thread at the top to make a vacuum tight seal. It will accept our 1/8 OD thermocouple probes. Lower portion of the adapter is reduced down to hold the thermocouple securely in place and away from the agitators. Supplied complete with a Celcon cap and Viton o-ring. Perfluoro o-rings are available.
Catalog No. CG1042E01 Supplier Chemglass Life Sciences Supplier No. CG1042E01
1/8" Thermocouple Adapter, 24/40 Inner. Adapter has a Chem-Thread at the top to make a vacuum tight seal. It will accept our 1/8 OD thermocouple probes. Lower portion of the adapter is reduced down to hold the thermocouple securely in place and away from the agitators. Supplied complete with a Celcon cap and Viton o-ring. Perfluoro o-rings are available.