Learn More
Chemglass Life Sciences Adapters, Inlet, 24/40 Inner Joint, 90° Swagelok, for 1/2" Compression Adapters
SDP
Catalog No. CG1014C01
Encompass_Preferred
Adapter, Inlet, Joint Size: 24/40, Dimensions Height x Width (MM): 70 x 50. Allows PTFE tubing to be connected to apparatus for introduction of vacuum or pressure. 1/2" OD ground swagelock is designed for use with a standard 1/2" TFE, PFA, or polypropylene compression fitting.
Catalog No. CG1014C01 Supplier Chemglass Life Sciences Supplier No. CG1014C01
Adapter, Inlet, Joint Size: 24/40, Dimensions Height x Width (MM): 70 x 50. Allows PTFE tubing to be connected to apparatus for introduction of vacuum or pressure. 1/2" OD ground swagelock is designed for use with a standard 1/2" TFE, PFA, or polypropylene compression fitting.