Learn More
Chemglass Life Sciences Adapters, Inlet, 24/40 Inner Joint, Straight Swagelok, for 1/2" Compression Adapters
SDP
Catalog No. CG1012C01
Encompass_Preferred
Adapter, inlet, inner joint, straight. Joint Size: 24/40, Overall Length (mm): 90. Allows PTFE tubing to be connected to apparatus for introduction of vacuum or pressure. 1/2" OD ground swagelock is designed for use with a standard 1/2" TFE, PFA, or polypropylene compression fitting.
Catalog No. CG1012C01 Supplier Chemglass Life Sciences Supplier No. CG1012C01
Adapter, inlet, inner joint, straight. Joint Size: 24/40, Overall Length (mm): 90. Allows PTFE tubing to be connected to apparatus for introduction of vacuum or pressure. 1/2" OD ground swagelock is designed for use with a standard 1/2" TFE, PFA, or polypropylene compression fitting.