Learn More
Chemglass Life Sciences Adapter, Outer Luer to 24/40 Inner
SDP
Catalog No. 50872876
Encompass_Preferred
Adapter, Outer Luer to 24/40 Inner. Adapter has an outer luer joint that provides a convenient way to connect solid phase extraction (SPE) cartridges, tubes or syringes to a standard taper inner joint. The adapter provides for easier sample concentration and clean-up while reducing the chances of cross-contamination. Hose connection is 10mm OD at the largest serration.
Catalog No. 50-872-876 Supplier Chemglass Life Sciences Supplier No. CG1049A24
Adapter, Outer Luer to 24/40 Inner. Adapter has an outer luer joint that provides a convenient way to connect solid phase extraction (SPE) cartridges, tubes or syringes to a standard taper inner joint. The adapter provides for easier sample concentration and clean-up while reducing the chances of cross-contamination. Hose connection is 10mm OD at the largest serration.